sec-Butylhydrazine dihydrochloride 95+%

Supplier: Apollo Scientific
Danger

OR318093-1G OR318093-25G OR318093-5G
APOSOR318093-1GEA 166 EUR
APOSOR318093-1G APOSOR318093-25G APOSOR318093-5G
sec-Butylhydrazine dihydrochloride 95+%
sec-Butylhydrazine dihydrochloride
Formula: C₄H₁₄Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD11840328
CAS Number: 1177361-36-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR