5-Bromo-7-nitro-1H-indole 95+%

Supplier: Apollo Scientific
Warning

OR318113-1G OR318113-5G
APOSOR318113-1GEA 78.3 EUR
APOSOR318113-1G APOSOR318113-5G
5-Bromo-7-nitro-1H-indole 95+%
5-Bromo-7-nitro-1H-indole
Formula: C₈H₅BrN₂O₂
MW: 241.05 g/mol
Melting Pt: 206…208 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00619260
CAS Number: 165669-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR