tert-Butyl piperazin-1-ylcarbamate 97%

Supplier: Apollo Scientific
Warning

OR321350-250MG
APOSOR321350-250MGEA 94.5 EUR
APOSOR321350-250MG
tert-Butyl piperazin-1-ylcarbamate 97%
tert-Butyl piperazin-1-ylcarbamate
Formula: C₉H₁₉N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11840387
CAS Number: 147081-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR