5-Chloroisothiazol-3-ol 97%

Supplier: Apollo Scientific
Warning

OR321363-1G
APOSOR321363-1GEA 145 EUR
APOSOR321363-1G
5-Chloroisothiazol-3-ol 97%
5-Chloroisothiazol-3-ol
Formula: C₃H₂ClNOS
Storage Temperature: Refrigerator
MDL Number: MFCD16658894
CAS Number: 25629-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR