tert-Butyl-4-acetylpiperidine-1-carboxylate 98%

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl 4-acetylpiperidine-1-carboxylate

OR321452-1G OR321452-5G OR321452-25G
APOSOR321452-1GEA 21.9 EUR
APOSOR321452-1G APOSOR321452-5G APOSOR321452-25G
tert-Butyl-4-acetylpiperidine-1-carboxylate 98%
tert-Butyl-4-acetylpiperidine-1-carboxylate
Formula: C₁₂H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD08437655
CAS Number: 206989-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR