5-Isopropyl-2-nitrobenzaldehyde

Supplier: Apollo Scientific

OR321525-1G OR321525-250MG
APOSOR321525-1GEA 348 EUR
APOSOR321525-1G APOSOR321525-250MG
5-Isopropyl-2-nitrobenzaldehyde
5-Isopropyl-2-nitrobenzaldehyde
Formula: C₁₀H₁₁NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD29120380
CAS Number: 1289211-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR