5-Methyl-2,4-diphenyl-2,3-dihydro-1H-pyrazol-3-one, tech

Supplier: Apollo Scientific

OR32181-1G
APOSOR32181-1GEA 1570 EUR
APOSOR32181-1G
5-Methyl-2,4-diphenyl-2,3-dihydro-1H-pyrazol-3-one, tech
5-Methyl-2,4-diphenyl-2,3-dihydro-1H-pyrazol-3-one
Formula: C₁₆H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD00140645
CAS Number: 79481-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR