5-Acetyl-2-(diethylamino)-6-methylpyridine-3-carbonitrile, tech

Supplier: Apollo Scientific

OR32203-1G
APOSOR32203-1GEA 1570 EUR
APOSOR32203-1G
5-Acetyl-2-(diethylamino)-6-methylpyridine-3-carbonitrile, tech
5-Acetyl-2-(diethylamino)-6-methylpyridine-3-carbonitrile
Formula: C₁₃H₁₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD00793586
CAS Number: 303146-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR