3-Chlorophenyl isocyanate

Supplier: Apollo Scientific
Danger

OR322031-1G OR322031-5G OR322031-10G
APOSOR322031-1GEA 26.1 EUR
APOSOR322031-1G APOSOR322031-5G APOSOR322031-10G
3-Chlorophenyl isocyanate
3-Chlorophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₄ClNO
Boiling Pt: 201 °C/760mmHg
Melting Pt: –4 °C
Density: 1,26 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002016
CAS Number: 2909-38-8
UN: 2206
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR