Ethyl 2-Bromoisovalerate 98%

Supplier: Apollo Scientific

Synonyms: 2-Bromoisovaleric Acid Ethyl Ester, 2-Bromo-3-methylbutyric Acid Ethyl Ester, Ethyl 2-bromo-3-methylbutanoate, Ethyl 2-Bromo-3-methylbutyrate

OR322033-25G
APOSOR322033-25GEA 56.7 EUR
APOSOR322033-25G
Ethyl 2-Bromoisovalerate 98%
Ethyl-2-bromoisovalerate
Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026855
CAS Number: 609-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR