1-(4-Nitrophenyl)-1H-pyrrole 95%

Supplier: Apollo Scientific

OR32237-25G
APOSOR32237-25GEA 126 EUR
APOSOR32237-25G
1-(4-Nitrophenyl)-1H-pyrrole 95%
1-(4-Nitrophenyl)-1H-pyrrole
Formula: C₁₀H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00119340
CAS Number: 4533-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR