(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt 97%

Supplier: Apollo Scientific
Danger

OR322596-1G OR322596-5G
APOSOR322596-1GEA 89.9 EUR
APOSOR322596-1G APOSOR322596-5G
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt 97%
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt
Formula: C₁₁H₁₇NO₄S
Density: 1,012 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066238
CAS Number: 104530-79-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR