(4-Aminophenyl)(4-methylpiperazin-1-yl)methanone

Supplier: Apollo Scientific
Warning

OR322867-1G
APOSOR322867-1GEA 155 EUR
APOSOR322867-1G
(4-Aminophenyl)(4-methylpiperazin-1-yl)methanone
(4-Aminophenyl)(4-methylpiperazin-1-yl)methanone
Formula: C₁₂H₁₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD00973892
CAS Number: 55121-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR