Methyl-2-amino-1H-benzo[d]imidazole-5-carboxylate

Supplier: Apollo Scientific

OR322965-250MG
APOSOR322965-250MGEA 130 EUR
APOSOR322965-250MG
Methyl-2-amino-1H-benzo[d]imidazole-5-carboxylate
Methyl-2-amino-1H-benzo[d]imidazole-5-carboxylate
Formula: C₉H₉N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD08752985
CAS Number: 106429-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR