2-Bromo-5H-pyrrolo[2,3-b]pyrazine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-4,7-diazaindole

OR323262-25G
APOSOR323262-25GEA 214 EUR
APOSOR323262-25G
2-Bromo-5H-pyrrolo[2,3-b]pyrazine 97%
2-Bromo-5H-pyrrolo[2,3-b]pyrazine
Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
Boiling Pt: 265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834822
CAS Number: 875781-43-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR