(3E)-3-[2-(2-Methylphenyl)hydrazin-1-ylidene]piperidin-2-one, tech

Supplier: Apollo Scientific

OR32419-1G
APOSOR32419-1GEA 1570 EUR
APOSOR32419-1G
(3E)-3-[2-(2-Methylphenyl)hydrazin-1-ylidene]piperidin-2-one, tech
(3E)-3-[2-(2-Methylphenyl)hydrazin-1-ylidene]piperidin-2-one
Formula: C₁₂H₁₅N₃O
Storage Temperature: Ambient
MDL Number: MFCD00794637
CAS Number: 14490-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR