1-(5-Chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)-1-ethanone 92%

Supplier: Apollo Scientific

OR32426-1G
APOSOR32426-1GEA 1230 EUR
APOSOR32426-1G
1-(5-Chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)-1-ethanone 92%
1-(5-Chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)-1-ethanone
Formula: C₁₂H₁₁ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD01314892
CAS Number: 321526-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR