1-Benzyl-3-[(E)-2-(furan-2-yl)ethenyl]urea, tech

Supplier: Apollo Scientific

OR32634-1G
APOSOR32634-1GEA 1570 EUR
APOSOR32634-1G
1-Benzyl-3-[(E)-2-(furan-2-yl)ethenyl]urea, tech
1-Benzyl-3-[(E)-2-(furan-2-yl)ethenyl]urea
Formula: C₁₄H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00794350
CAS Number: 478030-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR