(2E)-1-(3-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one 95%

Supplier: Apollo Scientific

OR32732-1G
APOSOR32732-1GEA 180 EUR
APOSOR32732-1G
(2E)-1-(3-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one 95%
(2E)-1-(3-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one
Formula: C₁₁H₁₂ClNO
MW: 209.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00172143
CAS Number: 876376-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR