Methyl-4-acetyl-1-methyl-1H-pyrrole-2-carboxylate 95%

Supplier: Apollo Scientific

OR32821-1G
APOSOR32821-1GEA 488 EUR
APOSOR32821-1G
Methyl-4-acetyl-1-methyl-1H-pyrrole-2-carboxylate 95%
Methyl-4-acetyl-1-methyl-1H-pyrrole-2-carboxylate
Formula: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00141903
CAS Number: 85795-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR