4-Chloro-2-(4-methylphenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one, tech

Supplier: Apollo Scientific

OR32872-1G
APOSOR32872-1GEA 1570 EUR
APOSOR32872-1G
4-Chloro-2-(4-methylphenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one, tech
4-Chloro-2-(4-methylphenyl)-5-[(prop-2-en-1-yl)amino]-2,3-dihydropyridazin-3-one
Formula: C₁₄H₁₄ClN₃O
MW: 275,733 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02083236
CAS Number: 478049-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR