Ethyl-4-hydroxy-8-methylquinoline-3-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Ethyl 8-methyl-4-oxo-1,4-dihydroquinoline-3-carboxylate

OR32980-5G
APOSOR32980-5GEA 81.2 EUR
APOSOR32980-5G
Ethyl-4-hydroxy-8-methylquinoline-3-carboxylate 95%
Ethyl 4-hydroxy-8-methylquinoline-3-carboxylate
Formula: C₁₃H₁₃NO₃
MW: 231.2500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173390
CAS Number: 77156-75-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR