4-Methyl-5,6,7,8-tetrahydroquinazolin-2-ol 95%

Supplier: Apollo Scientific

OR33003-1G
APOSOR33003-1GEA 336 EUR
APOSOR33003-1G
4-Methyl-5,6,7,8-tetrahydroquinazolin-2-ol 95%
4-Methyl-5,6,7,8-tetrahydroquinazolin-2-ol
Formula: C₉H₁₂N₂O
MW: 164,204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01444097
CAS Number: 88267-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR