4-(4-Chlorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile 95%

Supplier: Apollo Scientific

OR33061-1G OR33061-25G OR33061-5G
APOSOR33061-1GEA 180 EUR
APOSOR33061-1G APOSOR33061-25G APOSOR33061-5G
4-(4-Chlorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile 95%
4-(4-Chlorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile
Formula: C₁₁H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00099688
CAS Number: 39561-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR