Methyl-2-amino-2-(4-bromophenyl)acetate hydrochloride 95%

Supplier: Apollo Scientific

OR33179-1G OR33179-5G
APOSOR33179-1GEA 78.3 EUR
APOSOR33179-1G APOSOR33179-5G
Methyl-2-amino-2-(4-bromophenyl)acetate hydrochloride 95%
Methyl-2-amino-2-(4-bromophenyl)acetate hydrochloride
Formula: C₉H₁₁BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD05663802
CAS Number: 42718-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR