(1E)-4-(4-Chlorophenoxy)-1-(dimethylamino)pent-1-en-3-one, tech

Supplier: Apollo Scientific

OR33210-1G
APOSOR33210-1GEA 1570 EUR
APOSOR33210-1G
(1E)-4-(4-Chlorophenoxy)-1-(dimethylamino)pent-1-en-3-one, tech
(1E)-4-(4-Chlorophenoxy)-1-(dimethylamino)pent-1-en-3-one
Formula: C₁₃H₁₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00139114
CAS Number: 1164506-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR