4-Chloro-6-methyl-2-(propan-2-yl)pyrimidine 95%

Supplier: Apollo Scientific

Synonyms: 4-Chloro-2-isopropyl-6-methylpyrimidine

OR33282-1G
APOSOR33282-1GEA 671 EUR
APOSOR33282-1G
4-Chloro-6-methyl-2-(propan-2-yl)pyrimidine 95%
4-Chloro-6-methyl-2-(propan-2-yl)pyrimidine
Formula: C₈H₁₁ClN₂
MW: 170.639 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02685656
CAS Number: 4595-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR