4-Acetamidophenylacetonitrile

Supplier: Apollo Scientific
Warning

OR3333-1G OR3333-5G
APOSOR3333-1GEA 72.5 EUR
APOSOR3333-1G APOSOR3333-5G
4-Acetamidophenylacetonitrile
4-Acetamidophenylacetonitrile
Formula: C₁₀H₁₀N₂O
Melting Pt: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00016398
CAS Number: 25025-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR