N-{4-[2-(4-Acetamidophenyl)ethynyl]phenyl}acetamide, tech

Supplier: Apollo Scientific

OR33376-1G
APOSOR33376-1GEA 1570 EUR
APOSOR33376-1G
N-{4-[2-(4-Acetamidophenyl)ethynyl]phenyl}acetamide, tech
N-{4-[2-(4-Acetamidophenyl)ethynyl]phenyl}acetamide
Formula: C₁₈H₁₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01110979
CAS Number: 114879-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR