5-Bromo-2-hydrazinobenzoic acid

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2-carboxyphenylhydrazine

OR3382-500MG
APOSOR3382-500MGEA 203 EUR
APOSOR3382-500MG
5-Bromo-2-hydrazinobenzoic acid
5-Bromo-2-hydrazinobenzoic acid
Formula: C₇H₇BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD03094638
CAS Number: 177192-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR