3-Amino-2,4-dichloropyridine

Supplier: Apollo Scientific
Danger

OR345038-25G
APOSOR345038-25GEA 89.9 EUR
APOSOR345038-25G
3-Amino-2,4-dichloropyridine
3-Amino-2,4-dichloropyridine
Formula: C5H4Cl2N2
MW: 163.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750301
CAS Number: 173772-63-9
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR