3-Bromo-2-methoxybenzonitrile 98%

Supplier: Apollo Scientific

OR345437-25G
APOSOR345437-25GEA 136 EUR
APOSOR345437-25G
3-Bromo-2-methoxybenzonitrile 98%
3-Bromo-2-methoxybenzonitrile
Formula: C₈H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD09834773
CAS Number: 874472-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR