2-(1-Methyl-1H-pyrazol-3-yl)phenylamine

Supplier: Apollo Scientific
Warning

OR346099-500MG
APOSOR346099-500MGEA 0 EUR
APOSOR346099-500MG
2-(1-Methyl-1H-pyrazol-3-yl)phenylamine
2-(1-Methyl-1H-pyrazol-3-yl)phenylamine
Formula: C₁₀H₁₁N₃
MW: 173,22 g/mol
Melting Pt: 59…60 °C
Storage Temperature: Ambient
MDL Number: MFCD11052424
CAS Number: 87488-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR