2,5-Dichloro-pyridine-3-sulphonyl chloride

Supplier: Apollo Scientific

OR346212-1G OR346212-500MG OR346212-250MG
APOSOR346212-1GEA 405 EUR
APOSOR346212-1G APOSOR346212-500MG APOSOR346212-250MG
2,5-Dichloro-pyridine-3-sulphonyl chloride
2,5-Dichloro-pyridine-3-sulphonyl chloride
Formula: C₅H₂Cl₃NO₂S
Storage Temperature: Ambient
MDL Number: MFCD15142815
CAS Number: 1208081-36-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR