5-Chloro-3-phenyl-1,2,4-oxadiazole

Supplier: Apollo Scientific

OR346247-500MG
APOSOR346247-500MGEA 0 EUR
APOSOR346247-500MG
5-Chloro-3-phenyl-1,2,4-oxadiazole
5-Chloro-3-phenyl-1,2,4-oxadiazole
Formula: C₈H₅ClN₂O
MW: 180,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09971531
CAS Number: 827-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR