(4-Chlorocarbonylphenyl)boronic acid

Supplier: Apollo Scientific

OR360238-1G OR360238-25G OR360238-5G
APOSOR360238-1GEA 87 EUR
APOSOR360238-1G APOSOR360238-25G APOSOR360238-5G
(4-Chlorocarbonylphenyl)boronic acid
(4-Chlorocarbonylphenyl)boronic acid
Formula: C₇H₆BClO₃
Storage Temperature: Ambient
MDL Number: MFCD08062416
CAS Number: 332154-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR