6-Methoxypyridine-2-boronic acid

Supplier: Apollo Scientific
Warning

Synonyms: (6-Methoxypyridin-2-yl)boronic acid

OR360243-250MG OR360243-1G
APOSOR360243-250MGEA 109 EUR
APOSOR360243-250MG APOSOR360243-1G
6-Methoxypyridine-2-boronic acid
6-Methoxypyridine-2-boronic acid
Formula: C₆H₈BNO₃
MW: 152.944 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03093332
CAS Number: 372963-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR