(5-Cyano-2-methylphenyl)boronic acid 97%

Supplier: Apollo Scientific

OR360278-1G OR360278-5G
APOSOR360278-1GEA 56.7 EUR
APOSOR360278-1G APOSOR360278-5G
(5-Cyano-2-methylphenyl)boronic acid 97%
(5-Cyano-2-methylphenyl)boronic acid
Formula: C₈H₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD17015778
CAS Number: 867333-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR