3-Cyano-5-methylphenylboronic acid, pinacol ester 95+%

Supplier: Apollo Scientific

Synonyms: 3-Cyano-5-methylphenylboronic acid pinacol ester, 3-Methyl-5-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzonitrile

OR361035-1G
APOSOR361035-1GEA 81.2 EUR
APOSOR361035-1G
3-Cyano-5-methylphenylboronic acid, pinacol ester 95+%
3-Cyano-5-methylphenylboronic acid, pinacol ester
Formula: C₁₄H₁₈BNO₂
MW: 243.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383798
CAS Number: 1220219-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR