Iodomethylboronic acid, pinacol ester 95%

Supplier: Apollo Scientific
Warning

OR361281-5G OR361281-1G OR361281-10G
APOSOR361281-5GEA 30.6 EUR
APOSOR361281-5G APOSOR361281-1G APOSOR361281-10G
Iodomethylboronic acid, pinacol ester 95%
Iodomethylboronic acid, pinacol ester
Formula: C₇H₁₄BIO₂
Storage Temperature: Freezer
MDL Number: MFCD12405515
CAS Number: 70557-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR