4-(Methoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 97%

Supplier: Apollo Scientific

OR361328-250MG
APOSOR361328-250MGEA 55.1 EUR
APOSOR361328-250MG
4-(Methoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 97%
4-(Methoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester
Formula: C₁₄H₂₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD11520532
CAS Number: 151075-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR