3-(2-Methoxy-2-oxoethyl)phenylboronic acid 97%

Supplier: Apollo Scientific

OR361388-250MG
APOSOR361388-250MGEA 55.1 EUR
APOSOR361388-250MG
3-(2-Methoxy-2-oxoethyl)phenylboronic acid 97%
3-(2-Methoxy-2-oxoethyl)phenylboronic acid
Formula: C₉H₁₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD03425946
CAS Number: 643094-11-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR