2-Methoxyphenylboronic acid

Supplier: Apollo Scientific
Warning

OR3663-100G OR3663-25G OR3663-5G
APOSOR3663-100GEA 69.6 EUR
APOSOR3663-100G APOSOR3663-25G APOSOR3663-5G
2-Methoxyphenylboronic acid
2-Methoxyphenylboronic acid
Formula: C₇H₉BO₃
Melting Pt: 102…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236047
CAS Number: 5720-06-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR