7-Chloro-1H-indazole

Supplier: Apollo Scientific
Danger

OR3680-250MG
APOSOR3680-250MGEA 21.9 EUR
APOSOR3680-250MG
7-Chloro-1H-indazole
7-Chloro-1H-indazole
Formula: C₇H₅ClN₂
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD07368214
CAS Number: 37435-12-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR