2,3-Dichloro-N-(pyridin-3-ylmethyl)benzamide ≥98%

Supplier: Apollo Scientific

OR370076-1G
APOSOR370076-1GEA 545 EUR
APOSOR370076-1G
2,3-Dichloro-N-(pyridin-3-ylmethyl)benzamide ≥98%
2,3-Dichloro-N-(pyridin-3-ylmethyl)benzamide
Formula: C₁₃H₁₀Cl₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD02641047
CAS Number: 425685-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR