5-Bromo-2-chloro-4-(methylthio)pyrimidine 97%

Supplier: Apollo Scientific
Warning

OR3705-1G OR3705-5G
APOSOR3705-1GEA 45.1 EUR
APOSOR3705-1G APOSOR3705-5G
5-Bromo-2-chloro-4-(methylthio)pyrimidine 97%
5-Bromo-2-chloro-4-(methylthio)pyrimidine
Formula: C₅H₄BrClN₂S
Storage Temperature: Refrigerator
MDL Number: MFCD07781218
CAS Number: 59549-51-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR