4-(2-Bromobenzyl)morpholine

Supplier: Apollo Scientific
Warning

Synonyms: 4-(2-Bromophenylmethyl)morpholine, 1-Bromo-2-[(morpholin-4-yl)methyl]benzene

OR3761-1G
APOSOR3761-1GEA 60.9 EUR
APOSOR3761-1G
4-(2-Bromobenzyl)morpholine
4-(2-Bromobenzyl)morpholine
Formula: C₁₁H₁₄BrNO
Density: 1,4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00979294
CAS Number: 91130-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR