4-(4-Bromobenzyl)morpholine

Supplier: Apollo Scientific
Danger

OR3763-1G
APOSOR3763-1GEA 21.9 EUR
APOSOR3763-1G
4-(4-Bromobenzyl)morpholine
4-(4-Bromobenzyl)morpholine
Formula: C₁₁H₁₄BrNO
Melting Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00979535
CAS Number: 132833-51-3
UN: 3261
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR