α,4-Dichlorotoluene 99%

Supplier: Apollo Scientific
Danger

OR3764-1KG OR3764-250G OR3764-5KG OR3764-25G OR3764-100G OR3764-400G
APOSOR3764-1KGEA 104 EUR
APOSOR3764-1KG APOSOR3764-250G APOSOR3764-5KG APOSOR3764-25G APOSOR3764-100G APOSOR3764-400G
α,4-Dichlorotoluene 99%
α,4-Dichlorotoluene
Formula: C₇H₆Cl₂
Boiling Pt: 216…222 °C
Melting Pt: 26…30 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000914
CAS Number: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR