HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%

Supplier: Apollo Scientific
Danger

Synonyms: Hexamethyldisilazane

OR3890-250G OR3890-25G OR3890-1KG
APOSOR3890-250GEA 29 EUR
APOSOR3890-250G APOSOR3890-25G APOSOR3890-1KG
HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
Boiling Pt: 125…127 °C
Melting Pt: –80 °C
Density: 0,77 g/cm³ (25 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 3286
ADR: 3+6.1+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR